SY3H3D6823A8 Synthonix Corporation

4-(Trifluoromethyl)pyrimidin-5-amine

  • MDL number No MDL found
  • Cas Number : 1092299-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40791.html

Safety Information

MSDS https://synthonix.com/40791.html
Availability details for
Quantity
  • Availability
  • Add to Cart