SY3H3D6823A9 Synthonix Corporation

5-(Trifluoromethyl)pyridazin-3-amine

  • Cas Number : 1211591-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40792.html

Safety Information

MSDS https://synthonix.com/40792.html
Availability details for
Quantity
  • Availability
  • Add to Cart