SY3H3D6823AC Synthonix Corporation

Tetrazolo[1,5-a]pyridine

  • MDL number No MDL found
  • Cas Number : 274-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/40812.html

Safety Information

MSDS https://synthonix.com/40812.html
Availability details for
Quantity
  • Availability
  • Add to Cart