SY3H3D6823AF Synthonix Corporation

3H-[1,2,3]Triazolo[4,5-c]pyridine

  • Cas Number : 273-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40817.html

Safety Information

MSDS https://synthonix.com/40817.html
Availability details for
Quantity
  • Availability
  • Add to Cart