SY3H3D6823B2 Synthonix Corporation

[1,2,4]Triazolo[1,5-a]pyrazin-2-amine

  • MDL number No MDL found
  • Cas Number : 88002-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40987.html

Safety Information

MSDS https://synthonix.com/40987.html
Availability details for
Quantity
  • Availability
  • Add to Cart