SY3H3D6823B4 Synthonix Corporation

Thiophene-2-carbothioamide

  • MDL number MFCD00052890
  • Cas Number : 20300-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Pale-green - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41010.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41010.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart