SY3H3D6823BA Synthonix Corporation

2,4,6-Trichloro-3-nitropyridine

  • Cas Number : 60186-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41284.html

Safety Information

MSDS https://synthonix.com/41284.html
Availability details for
Quantity
  • Availability
  • Add to Cart