SY3H3D6823BC Synthonix Corporation

2,3,4,6-Tetrachloropyridine

  • MDL number No MDL found
  • Cas Number : 14121-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41297.html

Safety Information

MSDS https://synthonix.com/41297.html
Availability details for
Quantity
  • Availability
  • Add to Cart