SY3H3D6823BD Synthonix Corporation

2,4,5-Trichloro-6-(trifluoromethyl)pyrimidine

  • MDL number No MDL found
  • Cas Number : 84737-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41301.html

Safety Information

MSDS https://synthonix.com/41301.html
Availability details for
Quantity
  • Availability
  • Add to Cart