SY3H3D6823C6 Synthonix Corporation

1,3,5-Trifluoro-2,4,6-triiodobenzene

  • MDL number MFCD06248899
  • Cas Number : 84322-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41424.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41424.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart