SY3H3D6823D1 Synthonix Corporation

1,2,5-Trichloro-3-iodobenzene

  • MDL number No MDL found
  • Cas Number : 216393-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41657.html

Safety Information

MSDS https://synthonix.com/41657.html
Availability details for
Quantity
  • Availability
  • Add to Cart