SY3H3D6823D5 Synthonix Corporation

Thiophene-3,4-dicarbonitrile

  • Cas Number : 18853-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41697.html

Safety Information

MSDS https://synthonix.com/41697.html
Availability details for
Quantity
  • Availability
  • Add to Cart