SY3H3D6823D7 Synthonix Corporation

2,4,6-Trichloronicotinic acid

  • MDL number No MDL found
  • Cas Number : 69422-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41743.html

Safety Information

MSDS https://synthonix.com/41743.html
Availability details for
Quantity
  • Availability
  • Add to Cart