SY3H3D6823D8 Synthonix Corporation

2,5,6-Trichloronicotinic acid

  • MDL number No MDL found
  • Cas Number : 54718-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41744.html

Safety Information

MSDS https://synthonix.com/41744.html
Availability details for
Quantity
  • Availability
  • Add to Cart