SY3H3D6823DD Synthonix Corporation

2,3,5-Trichloroisonicotinaldehyde

  • MDL number No MDL found
  • Cas Number : 251997-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41759.html

Safety Information

MSDS https://synthonix.com/41759.html
Availability details for
Quantity
  • Availability
  • Add to Cart