SY3H3D682436 Synthonix Corporation

2-(Trifluoromethyl)pyrimidine-5-carbaldehyde

  • Cas Number : 304693-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41859.html

Safety Information

MSDS https://synthonix.com/41859.html
Availability details for
Quantity
  • Availability
  • Add to Cart