SY3H3D682439 Synthonix Corporation

2-(Trifluoromethyl)furan-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 536718-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42043.html

Safety Information

MSDS https://synthonix.com/42043.html
Availability details for
Quantity
  • Availability
  • Add to Cart