SY3H3D68243B Synthonix Corporation

2,4,5-Trifluorophenol

  • MDL number No MDL found
  • Cas Number : 2268-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42060.html

Safety Information

MSDS https://synthonix.com/42060.html
Availability details for
Quantity
  • Availability
  • Add to Cart