SY3H3D682443 Synthonix Corporation

2,3,5-Trichloroaniline

  • Cas Number : 18487-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42228.html

Safety Information

MSDS https://synthonix.com/42228.html
Availability details for
Quantity
  • Availability
  • Add to Cart