SY3H3D682444 Synthonix Corporation

2,3,6-Trichloroaniline

  • MDL number MFCD02683886
  • Cas Number : 88963-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42229.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42229.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart