SY3H3D682446 Synthonix Corporation

2,3,5-Trifluoroaniline

  • Cas Number : 363-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42246.html

Safety Information

MSDS https://synthonix.com/42246.html
Availability details for
Quantity
  • Availability
  • Add to Cart