SY3H3D682450 Synthonix Corporation

Thiophene-2,4-dicarbaldehyde

  • MDL number No MDL found
  • Cas Number : 932-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42325.html

Safety Information

MSDS https://synthonix.com/42325.html
Availability details for
Quantity
  • Availability
  • Add to Cart