SY3H3D682454 Synthonix Corporation

2,4,6-Trichloro-5-ethylpyrimidine

  • MDL number No MDL found
  • Cas Number : 1780-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42375.html

Safety Information

MSDS https://synthonix.com/42375.html
Availability details for
Quantity
  • Availability
  • Add to Cart