SY3H3D682455 Synthonix Corporation

3,4,5-Trifluorobenzene-1,2-diamine

  • MDL number No MDL found
  • Cas Number : 168966-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42376.html

Safety Information

MSDS https://synthonix.com/42376.html
Availability details for
Quantity
  • Availability
  • Add to Cart