SY3H3D682458 Synthonix Corporation

5-(Trifluoromethoxy)pyridin-2-amine

  • Cas Number : 1221171-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/42463.html

Safety Information

MSDS https://synthonix.com/42463.html
Availability details for
Quantity
  • Availability
  • Add to Cart