SY3H3D68245C Synthonix Corporation

2-(1H-Tetrazol-5-yl)pyridine

  • MDL number No MDL found
  • Cas Number : 33893-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42509.html

Safety Information

MSDS https://synthonix.com/42509.html
Availability details for
Quantity
  • Availability
  • Add to Cart