SY3H3D68245D Synthonix Corporation

(5-(Trifluoromethyl)pyridin-2-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1162257-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42602.html

Safety Information

MSDS https://synthonix.com/42602.html
Availability details for
Quantity
  • Availability
  • Add to Cart