SY3H3D68245E Synthonix Corporation

6H-Thieno[2,3-b]pyrrole

  • MDL number No MDL found
  • Cas Number : 250-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42680.html

Safety Information

MSDS https://synthonix.com/42680.html
Availability details for
Quantity
  • Availability
  • Add to Cart