SY3H3D682473 Synthonix Corporation

5,6,7,8-Tetrahydroimidazo[1,5-a]pyrazine hydrochloride

  • MDL number No MDL found
  • Cas Number : 601515-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43547.html

Safety Information

MSDS https://synthonix.com/43547.html
Availability details for
Quantity
  • Availability
  • Add to Cart