SY3H3D682474 Synthonix Corporation

5,6,7,8-Tetrahydroimidazo[1,2-a]pyrimidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1209264-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43548.html

Safety Information

MSDS https://synthonix.com/43548.html
Availability details for
Quantity
  • Availability
  • Add to Cart