SY3H3D682479 Synthonix Corporation

N,N,N-Trimethylpropan-1-aminium bromide

  • MDL number MFCD06227832
  • Cas Number : 2650-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43586.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43586.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart