SY3H3D682519 Synthonix Corporation

Trimethyl methanetricarboxylate

  • Cas Number : 1186-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43743.html

Safety Information

MSDS https://synthonix.com/43743.html
Availability details for
Quantity
  • Availability
  • Add to Cart