SY3H3D68251E Synthonix Corporation

trans-4-(Trifluoromethyl)cyclohexanol

  • Cas Number : 75091-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43787.html

Safety Information

MSDS https://synthonix.com/43787.html
Availability details for
Quantity
  • Availability
  • Add to Cart