SY3H3D682523 Synthonix Corporation

2,3,4-Trifluoro-5-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 197520-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44007.html

Safety Information

MSDS https://synthonix.com/44007.html
Availability details for
Quantity
  • Availability
  • Add to Cart