SY3H3D682526 Synthonix Corporation

2,5,6-Trichloro-4-methylnicotinonitrile

  • MDL number No MDL found
  • Cas Number : 63195-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44049.html

Safety Information

MSDS https://synthonix.com/44049.html
Availability details for
Quantity
  • Availability
  • Add to Cart