SY3H3D68252C Synthonix Corporation

6-(Trifluoromethyl)imidazo[2,1-b]thiazole-5-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 564443-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44081.html

Safety Information

MSDS https://synthonix.com/44081.html
Availability details for
Quantity
  • Availability
  • Add to Cart