SY3H3D68252E Synthonix Corporation

Thieno[3,2-b]thiophene-2-carbonitrile

  • Cas Number : 40985-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44190.html

Safety Information

MSDS https://synthonix.com/44190.html
Availability details for
Quantity
  • Availability
  • Add to Cart