SY3H3D682533 Synthonix Corporation

2,3,5-Trichlorobenzaldehyde

  • MDL number No MDL found
  • Cas Number : 56961-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44257.html

Safety Information

MSDS https://synthonix.com/44257.html
Availability details for
Quantity
  • Availability
  • Add to Cart