SY3H3D68253A Synthonix Corporation

5-(Thiazol-2-yl)isoxazole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 933759-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44374.html

Safety Information

MSDS https://synthonix.com/44374.html
Availability details for
Quantity
  • Availability
  • Add to Cart