SY3H3D68254D Synthonix Corporation

(2,3,4,5-Tetrafluorophenyl)methanol

  • MDL number No MDL found
  • Cas Number : 53072-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44684.html

Safety Information

MSDS https://synthonix.com/44684.html
Availability details for
Quantity
  • Availability
  • Add to Cart