SY3H3D682553 Synthonix Corporation

1,2,3-Trichloro-4-methylbenzene

  • MDL number No MDL found
  • Cas Number : 7359-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44730.html

Safety Information

MSDS https://synthonix.com/44730.html
Availability details for
Quantity
  • Availability
  • Add to Cart