SY3H3D68255B Synthonix Corporation

((Tribromomethyl)sulfonyl)benzene

  • MDL number No MDL found
  • Cas Number : 17025-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/45085.html

Safety Information

MSDS https://synthonix.com/45085.html
Availability details for
Quantity
  • Availability
  • Add to Cart