SY3H3D682560 Synthonix Corporation

3,4,5-Trichloro-2,6-dimethylpyridine

  • MDL number No MDL found
  • Cas Number : 28597-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/45321.html

Safety Information

MSDS https://synthonix.com/45321.html
Availability details for
Quantity
  • Availability
  • Add to Cart