SY3H3D682564 Synthonix Corporation

(3,4,5-Trifluorophenyl)methanamine

  • Cas Number : 235088-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45324.html

Safety Information

MSDS https://synthonix.com/45324.html
Availability details for
Quantity
  • Availability
  • Add to Cart