SY3H3D68269F Synthonix Corporation

3-(Trifluoromethyl)benzene-1,2-diamine

  • MDL number No MDL found
  • Cas Number : 360-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/45624.html

Safety Information

MSDS https://synthonix.com/45624.html
Availability details for
Quantity
  • Availability
  • Add to Cart