SY3H3D6826A9 Synthonix Corporation

4-Thioureidobenzenesulfonamide

  • Cas Number : 1718-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46351.html

Safety Information

MSDS https://synthonix.com/46351.html
Availability details for
Quantity
  • Availability
  • Add to Cart