SY3H3D6826AA Synthonix Corporation

p-Tolylphosphonic acid

  • MDL number No MDL found
  • Cas Number : 3366-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/46447.html

Safety Information

MSDS https://synthonix.com/46447.html
Availability details for
Quantity
  • Availability
  • Add to Cart