SY3H3D6826AB Synthonix Corporation

2-(Tetrahydro-2H-pyran-4-yl)ethanamine hydrochloride

  • Cas Number : 389621-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46515.html

Safety Information

MSDS https://synthonix.com/46515.html
Availability details for
Quantity
  • Availability
  • Add to Cart