SY3H3D6826AC Synthonix Corporation

2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone hydrochloride

  • MDL number No MDL found
  • Cas Number : 1588441-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46526.html

Safety Information

MSDS https://synthonix.com/46526.html
Availability details for
Quantity
  • Availability
  • Add to Cart