SY3H3D6826B2 Synthonix Corporation

(2-(Trifluoromethoxy)phenyl)hydrazine hydrochloride

  • MDL number No MDL found
  • Cas Number : 133115-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46540.html

Safety Information

MSDS https://synthonix.com/46540.html
Availability details for
Quantity
  • Availability
  • Add to Cart